N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol
製品番号(カタログ番号) A12033.18
または、製品番号A12033-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25561-30-2
IUPAC Nametrimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular FormulaC8H18F3NOSi2
InChI KeyXCOBLONWWXQEBS-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
さらに表示
Assay (GC)≥98.0%
Refractive Index1.3820-1.3860 @ 20°C (non-U.S. specification)
FormLiquid
Identification (FTIR)Conforms (non-U.S. specification)
Appearance (Color)Clear colorless to pale yellow
さらに表示
N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

Solubility
Miscible with chloroform.

Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Powerful silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions, similar to N,O-Bis(trimethyl silyl) acetamide, L00183, with the advantage that both the reagent and its by-product, trifluoroacetamide, are more volatile than the corresponding non-fluorinated analogues: Biochem. Biophys. Res. Commun., 31, 616 (1968). See Appendix 4.
  2. Sagona, J. A.; Dukett, J. E.; Hawley, H. A.; Mazurek, M. A. Sequential derivatization of polar organic compounds in cloud water using O-(2, 3, 4, 5, 6-pentafluorobenzyl) hydroxylamine hydrochloride, N, O-bis (trimethylsilyl) trifluoroacetamide, and gas-chromatography/mass spectrometry analysis. J. Chromatogr. A 2014, 1362, 16-24.
  3. Irei, S.; Rudolph, J.; Huang, L. Compound-specific stable carbon isotope ratios of phenols and nitrophenols derivatized with N, O-bis (trimethylsilyl) trifluoroacetamide. Anal. Chim. Acta 2013, 786, 95-102.