Thermo Scientific Chemicals

L-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A12111.36
500 g, Each
Thermo Scientific Chemicals

L-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A12111.36
500 g, Each
Quantity
Catalog number: A12111.36
also known as A12111-36
Price (USD)
Price: 292.00
Online price: 248.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3184-13-2
IUPAC Name
hydrogen 2,5-diaminopentanoic acid chloride
Molecular Formula
C5H13ClN2O2
InChI Key
GGTYBZJRPHEQDG-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NCCCC(N)C(O)=O
Heavy Metals
≤10ppm (US sourced material)
Identification (FTIR)
Conforms (UK sourced material)
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (UK sourced material)
Appearance (Color)
White

Description

L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.

Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.

Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text