Thermo Scientific Chemicals

Dimethylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A12133.30
250 g, Each
Thermo Scientific Chemicals

Dimethylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A12133.30
250 g, Each
Quantity

Chemical Identifiers

CAS
506-59-2
IUPAC Name
hydrogen dimethylamine chloride
Molecular Formula
C2H8ClN
InChI Key
IQDGSYLLQPDQDV-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CNC
Water Content (Karl Fischer Titration)
≤2.0%
Appearance (Color)
White to pale yellow
Form
Crystals or compacted crystalline powder or crystalline solid
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms

Description

Dimethylamine hydrochloride is used as an intermediate in the manufacture of pharmaceuticals like ranitidine and metformin, tramadol and amlodipine. It is used as a precursor of atrazine. It is associated with sodium acetate and used to carry out the Willgerodt- Kindler reaction to prepare amides. Its free base reacts with carbon disulfide to get dimethyldithiocarbamate which is used in rubber vulcanization. It is involved in the synthesis of dimethyl-(1-methyl-pyrrol-2-ylmethyl)-amine by reacting with 1-methyl pyrrole and formaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylamine hydrochloride is used as an intermediate in the manufacture of pharmaceuticals like ranitidine and metformin, tramadol and amlodipine. It is used as a precursor of atrazine. It is associated with sodium acetate and used to carry out the Willgerodt- Kindler reaction to prepare amides. Its free base reacts with carbon disulfide to get dimethyldithiocarbamate which is used in rubber vulcanization. It is involved in the synthesis of dimethyl-(1-methyl-pyrrol-2-ylmethyl)-amine by reacting with 1-methyl pyrrole and formaldehyde.

Solubility
Soluble in water, alcohol and chloroform

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text