N-(Benzyloxycarbonyloxy)succinimide, 98%
N-(Benzyloxycarbonyloxy)succinimide, 98%
N-(Benzyloxycarbonyloxy)succinimide, 98%
Thermo Scientific Chemicals

N-(Benzyloxycarbonyloxy)succinimide, 98%

CAS: 13139-17-8 | C12H11NO5 | 249.222 g/mol
製品番号(カタログ番号) A12153.22
または、製品番号A12153-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13139-17-8
IUPAC Namebenzyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular FormulaC12H11NO5
InChI KeyMJSHDCCLFGOEIK-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)ON1C(=O)CCC1=O
さらに表示
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Melting Point (clear melt)77.0-83.0?C
Appearance (Color)White
Identification (FTIR)Conforms
N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

Solubility
Soluble in chloroform, methanol, acetone (0.1 g/mL).

Notes
Moisture sensitive. Storage temperature 2-8°C. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Hassan Ali Zamani. Erbium(III) PVC membrane sensor based on N-(benzyloxycarbonyloxy)succinimide as a new neutral ionophore. Chin. Chem. Lett. 2011, 22 (3), 346-349.
  2. Selective reagent for the introduction of the benzyloxycarbonyl (Cbz or Z) protecting group, avoiding the formation of amino acid dimers: Tetrahedron Lett., 4765 (1966); J. Antibiot., 25, 695 (1972); 34, 513 (1981); Can. J. Chem., 60, 976 (1982).
  3. Especially convenient for the selective protection of the -amino group of lysine and ornithine: Can. J. Chem., 54, 733 (1976); Rec. Trav. Chim., 99, 400 (1980). See Appendix 6.