Glycine hydrochloride, 98%
Glycine hydrochloride, 98%
Glycine hydrochloride, 98%
Glycine hydrochloride, 98%
Thermo Scientific Chemicals

Glycine hydrochloride, 98%

CAS: 6000-43-7 | C2H6ClNO2 | 111.53 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A12162.22
または、製品番号A12162-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6000-43-7
IUPAC Namehydrogen 2-aminoacetic acid chloride
Molecular FormulaC2H6ClNO2
InChI KeyIVLXQGJVBGMLRR-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(O)=O
さらに表示
Appearance (Color)White to pale cream
FormCrystalline powder
Assay from Suppliers CofA≥97.5%
Melting Point178-182°C
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Soluble in water (>100 g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Michael G. Wohl; M.D.; Nathan Pastor, M.D. Adiposis Dolorosa (Dercum's Disease) Treatment Of The Asthenic Phase With Prostigmine And Aminoacetic Acid. Jama. 1938, 110 (16), 1261-1264.
  2. George L. Maison, M.D. Failure Of Gelatin Or Amino-Acetic Acid To Increase The Work Ability Of Individual Normal Human Muscles. Tetrahedron Letters. 1940, 115 (17), 1439-1441.