2-Chlorobenzimidazole, 97%
2-Chlorobenzimidazole, 97%
2-Chlorobenzimidazole, 97%
Thermo Scientific Chemicals

2-Chlorobenzimidazole, 97%

CAS: 4857-06-1 | C7H5ClN2 | 152.58 g/mol
製品番号(カタログ番号) A12193.14
または、製品番号A12193-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4857-06-1
IUPAC Name2-chloro-1H-1,3-benzodiazole
Molecular FormulaC7H5ClN2
InChI KeyAYPSHJCKSDNETA-UHFFFAOYSA-N
SMILESClC1=NC2=CC=CC=C2N1
さらに表示
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.
RUO – Research Use Only

General References:

  1. Ryuichi Iemura; Tsuneo Kawashima; Toshikazu Fukuda; Keizo Ito; Goro Tsukamoto. Synthesis of 2-(4-substituted-1-piperazinyl)benzimidazoles as H1-antihistaminic agents. J. Med. Chem. 1986, 29 (7), 1178-1183.
  2. D. Harrison; J. T. Ralph and A. C. B. Smith. Some 1- and 2-halogenobenzimidazoles. J. Chem. Soc. 1963, 2930-2937.