1,3-Indanedione, 97%
1,3-Indanedione, 97%
1,3-Indanedione, 97%
Thermo Scientific Chemicals

1,3-Indanedione, 97%

CAS: 606-23-5 | C9H6O2 | 146.145 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A12206.22
または、製品番号A12206-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS606-23-5
IUPAC Name2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O2
InChI KeyUHKAJLSKXBADFT-UHFFFAOYSA-N
SMILESO=C1CC(=O)C2=CC=CC=C12
さらに表示
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)124-131?C
Appearance (Color)White to cream or yellow to yellow-green to pale green to pale brown
FormPowder or crystals
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the first stage of forensic identification of latent fingerprints. It is particularly useful for paper, and for items printed with thermal inks such as receipts.

Solubility
Soluble in ethanol, ether, benzene, slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Sook-Hee Kim; Seong-Jin Hong; Jaeduk Yoo; Sung Kuk Kim; Janathan L. Sessler and Chang-Hee Lee. Strapped Calix[4]pyrroles Bearing a 1,3-Indanedione at a β-Pyrrolic Position: Chemodosimeters for the Cyanide Anion. Org. Lett. 2009, 11 (16), 3626-3629.
  2. Nalan Tekin; Hilmi Namli; Onur Turhan. Solvents effect on infrared spectra of 1,3-indanedione in organic solvents. Vibrational Spectroscopy. 2005, 39 (2), 214-219.
  3. The oxidative conversion of ß-diketones to carboxylic acids with cerium(IV) ammonium nitrate is illustrated by the high-yield conversion of 1,3-indanedione to phthalic anhydride at room temperature in acetonitrile: J. Org. Chem., 71, 4516 (2006).