Thermo Scientific Chemicals

Phthalazine, 98%, Thermo Scientific Chemicals

Catalog number: A12270.06
5 g, Each
Thermo Scientific Chemicals

Phthalazine, 98%, Thermo Scientific Chemicals

Catalog number: A12270.06
5 g, Each
Quantity
Catalog number: A12270.06
also known as A12270-06
Price (USD)
Price: 76.70
Online price: 65.65
Your price:
Quantity
-

Chemical Identifiers

CAS
253-52-1
IUPAC Name
phthalazine
Molecular Formula
C8H6N2
InChI Key
LFSXCDWNBUNEEM-UHFFFAOYSA-N
SMILES
C1=CC=C2C=NN=CC2=C1
Melting Point (clear melt)
86.0-93.0?C
Appearance (Color)
Yellow to brown
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid

Description

Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalazine derivatives including copper and platinum complexes were induce the apoptosis in breast cancer cell lines and renal hyperemia.

Solubility
Soluble in water, methanol, chloroform.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text