4-Chlorobenzoylacetonitrile, 98%
4-Chlorobenzoylacetonitrile, 98%
4-Chlorobenzoylacetonitrile, 98%
Thermo Scientific Chemicals

4-Chlorobenzoylacetonitrile, 98%

CAS: 4640-66-8 | C9H6ClNO | 179.603 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A12364.06
または、製品番号A12364-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4640-66-8
IUPAC Name3-(4-chlorophenyl)-3-oxopropanenitrile
Molecular FormulaC9H6ClNO
InChI KeyJYOUFPNYTOFCSJ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)CC#N
さらに表示
Melting Point (clear melt)126.0-133.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to yellow to orange
Assay (GC)≥97.5%
Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Po Huang Liang,; Guang Yang,; Debra Dunaway-Mariano.Specificity of 4-chlorobenzoyl coenzyme A dehalogenase catalyzed dehalogenation of halogenated aromatics. Biochemistry. 1993, 32(45),12245-12250.
  2. Gwen P. Crooks,; Shelley D. Copley. Purification and Characterization of 4-Chlorobenzoyl CoA Dehalogenase from Arthrobacter sp. Strain 4-CB1. Biochemistry. 1994. 33 (38),11645-11649.