Thermo Scientific Chemicals

Butyric anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A12377.AP
500 mL, Each
Thermo Scientific Chemicals

Butyric anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A12377.AP
500 mL, Each
Quantity
Catalog number: A12377.AP
also known as A12377-AP
Price (USD)
Price: 45.50
Online price: 43.65
Your price:
Quantity
-

Chemical Identifiers

CAS
106-31-0
IUPAC Name
butanoyl butanoate
Molecular Formula
C8H14O3
InChI Key
YHASWHZGWUONAO-UHFFFAOYSA-N
SMILES
CCCC(=O)OC(=O)CCC
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4115-1.4155 @ 20?C

Description

Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyric anhydride is used in the preparation of amidoamine dendron-based co-adsorbents, which finds application in dye- sensitized solar cells improvement. It is also used in the synthesis of butyrate ester, which is used as a perfume and flavor. Further, it acts as a fumigant to drive bees from their hives. In addition to this, it is used in food additives, textile auxiliaries, varnishes, perfumes, pharmaceuticals and disinfectants.

Solubility
Miscible with water and ether.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text