Thermo Scientific Chemicals

L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A12386.14
25 g, Each
Thermo Scientific Chemicals

L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A12386.14
25 g, Each
Quantity
Catalog number: A12386.14
also known as A12386-14
Price (USD)
Price: 80.20
Online price: 68.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7389-87-9
IUPAC Name
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl
Molecular Formula
C7H11ClN3O2
InChI Key
VXXIPPPPKNWFLK-UHFFFAOYNA-N
SMILES
[Cl].COC(=O)C(N)CC1=CN=CN1
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Appearance (Color)
White
Optical Rotation
10.0 ± 1.0? (C=2 in water)

Description

L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text