S-Methyl thioacetate, 98+%
S-Methyl thioacetate, 98+%
S-Methyl thioacetate, 98+%
S-Methyl thioacetate, 98+%
Thermo Scientific Chemicals

S-Methyl thioacetate, 98+%

CAS: 1534-08-3 | C3H6OS | 90.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12400.06
also known as A12400-06
Price (USD)
72.65
Online Exclusive
80.60
Save 7.95 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.60
Save 7.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS1534-08-3
IUPAC Name1-(methylsulfanyl)ethan-1-one
Molecular FormulaC3H6OS
InChI KeyOATSQCXMYKYFQO-UHFFFAOYSA-N
SMILESCSC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.4615-1.4655 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only