Thermo Scientific Chemicals

Isatin, 98%, Thermo Scientific Chemicals

Catalog number: A12468.0E
2500 g, Each
Thermo Scientific Chemicals

Isatin, 98%, Thermo Scientific Chemicals

Catalog number: A12468.0E
2500 g, Each
Quantity
Catalog number: A12468.0E
also known as A12468-0E
Price (USD)
Price: 422.00
Online price: 333.65
Your price:
Quantity
-

Chemical Identifiers

CAS
91-56-5
IUPAC Name
2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C8H5NO2
InChI Key
JXDYKVIHCLTXOP-UHFFFAOYSA-N
SMILES
O=C1NC2=CC=CC=C2C1=O
Water Content (Karl Fischer Titration)
≤1%
Appearance (Color)
Orange
Form
Powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

Isatin is an endogenous monoamine oxidize (MAO) inhibitor involved in stress and anxiety. Additionally, isatin inhibits alkaline phosphatase (ALP), nitric oxide (NO)-stimulated soluble guanylate cyclase, and other enzymes. Isatins undergo a one-pot Wolff-Kishner like reduction with hydrazine hydrate, under surprisingly mild conditions, to give the corresponding oxindoles. It is used as chromatographic spray reagent for amino acid detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isatin is an endogenous monoamine oxidize (MAO) inhibitor involved in stress and anxiety. Additionally, isatin inhibits alkaline phosphatase (ALP), nitric oxide (NO)-stimulated soluble guanylate cyclase, and other enzymes. Isatins undergo a one-pot Wolff-Kishner like reduction with hydrazine hydrate, under surprisingly mild conditions, to give the corresponding oxindoles. It is used as chromatographic spray reagent for amino acid detection.

Solubility
Soluble in water (1.9 g/L at 20°C).

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text