Tris(4-chlorophenyl)phosphine, 97%
Tris(4-chlorophenyl)phosphine, 97%
Tris(4-chlorophenyl)phosphine, 97%
Tris(4-chlorophenyl)phosphine, 97%
Thermo Scientific Chemicals

Tris(4-chlorophenyl)phosphine, 97%

CAS: 1159-54-2 | C18H12Cl3P | 365.618 g/mol
製品番号(カタログ番号) A12487.06
または、製品番号A12487-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1159-54-2
IUPAC Nametris(4-chlorophenyl)phosphane
Molecular FormulaC18H12Cl3P
InChI KeyIQKSLJOIKWOGIZ-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)P(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1
さらに表示
Melting Point96-103°C
Appearance (Color)White to cream or pale yellow
FormCrystalline powder or powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only