5-Chloro-2-nitrobenzoic acid, 99%
5-Chloro-2-nitrobenzoic acid, 99%
5-Chloro-2-nitrobenzoic acid, 99%
Thermo Scientific Chemicals

5-Chloro-2-nitrobenzoic acid, 99%

CAS: 2516-95-2 | C7H4ClNO4 | 201.562 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A12493.06
または、製品番号A12493-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2516-95-2
IUPAC Name5-chloro-2-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyZKUYSJHXBFFGPU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Cl)=CC=C1[N+]([O-])=O
さらに表示
Assay (Aqueous acid-base Titration)>98.5 to <101.5%
Appearance (Color)White to pale cream or pale yellow to pale brown
FormPowder
Assay (Silylated GC)>98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only