3-Fluorobenzyl alcohol, 98%
3-Fluorobenzyl alcohol, 98%
3-Fluorobenzyl alcohol, 98%
3-Fluorobenzyl alcohol, 98%
Thermo Scientific Chemicals

3-Fluorobenzyl alcohol, 98%

CAS: 456-47-3 | C7H7FO | 126.13 g/mol
数量:
10 g
25 g
50 g
製品番号(カタログ番号) A12509.18
または、製品番号A12509-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS456-47-3
IUPAC Name(3-fluorophenyl)methanol
Molecular FormulaC7H7FO
InChI KeyQDHRSLFSDGCJFX-UHFFFAOYSA-N
SMILESOCC1=CC=CC(F)=C1
さらに表示
Appearance (Color)Clear colorless to yellow or pale orange
Refractive Index1.5080-1.5120 @ 20?C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only