Ethyl crotonate, 98%
Ethyl crotonate, 98%
Ethyl crotonate, 98%
Thermo Scientific Chemicals

Ethyl crotonate, 98%

CAS: 623-70-1 | C6H10O2 | 114.14 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A12529.22
または、製品番号A12529-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS623-70-1
IUPAC Nameethyl (2E)-but-2-enoate
Molecular FormulaC6H10O2
InChI KeyZFDIRQKJPRINOQ-HWKANZROSA-N
SMILESCCOC(=O)\C=C\C
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4220-1.4260 @ 20°C
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J.L.Herrmann; G.R.Kieczykowski; R.H.Schlessinger. Deconjugative alkylation of the enolate anion derived from ethyl crotonate. Tetrahedron Letters. 1973, 14, (26)2433-2436
  2. Jerry R.Mohrig; Sabrina S.Fu; Randall W.King; Ronald Warnet; Gary Gustafson. Stereochemistry of nucleophilic conjugate addition. Addition of ethanol-d and 2-methyl-2-propanethiol-d to ethyl crotonate. J. Am. Chem. Soc. 1990, 112, (9)3665-3667