Thermo Scientific Chemicals

4-n-Hexylresorcinol, 99%, Thermo Scientific Chemicals

Catalog number: A12552.14
25 g, Each
Thermo Scientific Chemicals

4-n-Hexylresorcinol, 99%, Thermo Scientific Chemicals

Catalog number: A12552.14
25 g, Each
Quantity
Catalog number: A12552.14
also known as A12552-14
Price (USD)
Price: 63.20
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
136-77-6
Melting Point (clear melt)
63.5-69.5?C
Appearance (Color)
White to pale brown
Form
Powder
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

4-Hexylresorcinol is an inhibitor of transglutaminase-2 in KB cells. Studies indicate that 4-Hexylresorcinol reduces DNA oxidative damage in human lymphocytes. is a substituted dihydroxybenzene used topically as an antiseptic for the treatment of minor skin infections. is a substituted phenol with bactericidal, antihelminthic and potential antineoplastic activities. The combination of 4-Hexylresorcinol and Cisplatin has been shown to inhibit tumor growth, cell growth, and KB cell viability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexylresorcinol is an inhibitor of transglutaminase-2 in KB cells. Studies indicate that 4-Hexylresorcinol reduces DNA oxidative damage in human lymphocytes. is a substituted dihydroxybenzene used topically as an antiseptic for the treatment of minor skin infections. is a substituted phenol with bactericidal, antihelminthic and potential antineoplastic activities. The combination of 4-Hexylresorcinol and Cisplatin has been shown to inhibit tumor growth, cell growth, and KB cell viability.

Solubility
Soluble in water. 0.05 g/100 mL (18°C)

Notes
Incompatible with acid chlorides, acid anhydrides, oxidizing agents.  Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text