Thermo Scientific Chemicals

Cyclopropanemethanol, 98%, Thermo Scientific Chemicals

Catalog number: A12567.14
25 g, Each
Thermo Scientific Chemicals

Cyclopropanemethanol, 98%, Thermo Scientific Chemicals

Catalog number: A12567.14
25 g, Each
Quantity
Catalog number: A12567.14
also known as A12567-14
Price (USD)
Price: 72.40
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2516-33-8
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4310-1.4350 @ 20?C
Form
Liquid

Description

Cyclopropanemethanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethanol is used as an organic chemical synthesis intermediate.

Solubility
Fully miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text