DL-Alanine methyl ester hydrochloride, 98+%
DL-Alanine methyl ester hydrochloride, 98+%
DL-Alanine methyl ester hydrochloride, 98+%
DL-Alanine methyl ester hydrochloride, 98+%
Thermo Scientific Chemicals

DL-Alanine methyl ester hydrochloride, 98+%

CAS: 13515-97-4 | C4H10ClNO2 | 139.58 g/mol
製品番号(カタログ番号) A12599.06
または、製品番号A12599-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13515-97-4
IUPAC Namemethyl 2-aminopropanoate hydrochloride
Molecular FormulaC4H10ClNO2
InChI KeyIYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILESCl.COC(=O)C(C)N
さらに表示
FormPowder or lumps
Appearance (Color)White to pale yellow
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only