Thermo Scientific Chemicals

4-Hydroxyquinoline-2-carboxylic acid hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A12602.14
25 g, Each
Thermo Scientific Chemicals

4-Hydroxyquinoline-2-carboxylic acid hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A12602.14
25 g, Each
Quantity
Catalog number: A12602.14
also known as A12602-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
345909-35-5
IUPAC Name
4-hydroxyquinoline-2-carboxylic acid
Molecular Formula
C10H7NO3
InChI Key
HCZHHEIFKROPDY-UHFFFAOYSA-N
SMILES
OC(=O)C1=NC2=CC=CC=C2C(O)=C1
Form
Powder
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
4.54-12.5% (0.5-1.5 waters)
Appearance (Color)
Dark cream to cream to yellow to green to pale brown to pale gray to gray

Description

Broad spectrum excitatory amino acid antagonist4-Hydroxyquinoline-2-carboxylic acid hydrate is used as an intermediate in synthetic chemistry. It acts as an antiexcitotoxic and anticonvulsant. It finds application in certain neurobiological disorders therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot ethanol and water. Insoluble in ether. 

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text