2-Bromo-4'-chloroacetophenone, 98%
2-Bromo-4'-chloroacetophenone, 98%
2-Bromo-4'-chloroacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-4'-chloroacetophenone, 98%

CAS: 536-38-9 | C8H6BrClO | 233.489 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A12607.22
または、製品番号A12607-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS536-38-9
IUPAC Name2-bromo-1-(4-chlorophenyl)ethan-1-one
Molecular FormulaC8H6BrClO
InChI KeyFLAYZKKEOIAALB-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)CBr
さらに表示
Appearance (Color)White to cream
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)93.0-100.0?C
2-Bromo-4'-chloroacetophenone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. G.Madhuramba; P.Ramasamy; P.Anbusrinivasan; G.Vasudeva; S.Kavitha; S.Mojumdar. Growth and characterization studies of 2-bromo-4'-chloroacetophenone (BCAP) crystals. Journal of Thermal Analysis and Calorimetry. 2008, 94, (1)59-62