Triphenylvinylsilane, 98%
Triphenylvinylsilane, 98%
Triphenylvinylsilane, 98%
Thermo Scientific Chemicals

Triphenylvinylsilane, 98%

CAS: 18666-68-7 | C20H18Si | 286.449 g/mol
製品番号(カタログ番号) A12713.14
または、製品番号A12713-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン913423
CAS18666-68-7
化学物質名または材質Triphenylvinylsilane
EINECS番号242-487-4
式量286.45
さらに表示
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture and heat. Store under inert gas.
RUO – Research Use Only

General References:

  1. Yasuyuki Ura.; Ryuichiro Hara.; Tamotsu Takahashi. Preparation of ethylene-bridged Group 14 metal-zirconocene complexes.J. Organomet. Chem. 2000,611(1-2), 299-303.
  2. Louis F. Cason, Houston G. Brooks. The Addition Of Organolithium Compounds To Vinyltriphenyl- And Vinyltrimethyl-Silanes.J. Org. Chem.1954,19(8), 1278-1282.