Anthranilic acid, 98+%
Thermo Scientific Chemicals

Anthranilic acid, 98+%

数量:
250 g
1000 g
5000 g
製品番号(カタログ番号) A12715.0I
または、製品番号A12715-0I
価格(JPY)
-
数量:
5000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS118-92-3
IUPAC Name2-aminobenzoic acid
Molecular FormulaC7H7NO2
InChI KeyRWZYAGGXGHYGMB-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C(O)=O
さらに表示
Assay from Supplier's CofA≥98.0% (HPLC)
Appearance (Color)Pale yellow or cream to pale brown
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Melting Point (clear melt)144-148?C
Anthranilic acid can be used in organic synthesis to generate the benzyne intermediate. It is used as an intermediate for production of dyes, pigments, and saccharin. It and its esters are used in preparing perfumes to imitate jasmine and orange, pharmaceuticals and UV-absorber as well as corrosion inhibitors for metals and mold inhibitors in soya sauce.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.