Thermo Scientific Chemicals

Anthranilic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12715.30
250 g, Each
Thermo Scientific Chemicals

Anthranilic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12715.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A12715.30
also known as A12715-30
Price (USD)
Price: 34.10
Your price:
Quantity
-

Chemical Identifiers

CAS
118-92-3
IUPAC Name
2-aminobenzoic acid
Molecular Formula
C7H7NO2
InChI Key
RWZYAGGXGHYGMB-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1C(O)=O
Assay from Supplier's CofA
≥98.0% (HPLC)
Appearance (Color)
Pale yellow or cream to pale brown
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
144-148?C

Description

Anthranilic acid can be used in organic synthesis to generate the benzyne intermediate. It is used as an intermediate for production of dyes, pigments, and saccharin. It and its esters are used in preparing perfumes to imitate jasmine and orange, pharmaceuticals and UV-absorber as well as corrosion inhibitors for metals and mold inhibitors in soya sauce.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text