3,4,5-Trimethoxycinnamic acid, predominantly trans, 99%
3,4,5-Trimethoxycinnamic acid, predominantly trans, 99%
3,4,5-Trimethoxycinnamic acid, predominantly trans, 99%
Thermo Scientific Chemicals

3,4,5-Trimethoxycinnamic acid, predominantly trans, 99%

CAS: 90-50-6 | C12H14O5 | 238.239 g/mol
製品番号(カタログ番号) A12767.14
または、製品番号A12767-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS90-50-6
IUPAC Name(2E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
Molecular FormulaC12H14O5
InChI KeyYTFVRYKNXDADBI-SNAWJCMRSA-N
SMILESCOC1=CC(C=CC(O)=O)=CC(OC)=C1OC
さらに表示
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Assay (Silylated GC)≥98.5%
Melting Point (clear melt)125.0-131.0?C
FormPowder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only