Thermo Scientific Chemicals

Diethyl ketomalonate, 95%, Thermo Scientific Chemicals

Catalog number: A12771.06
5 g, Each
Thermo Scientific Chemicals

Diethyl ketomalonate, 95%, Thermo Scientific Chemicals

Catalog number: A12771.06
5 g, Each
Quantity
Catalog number: A12771.06
also known as A12771-06
Price (USD)
Price: 46.50
Online price: 39.65
Your price:
Quantity
-

Chemical Identifiers

CAS
609-09-6
IUPAC Name
1,3-diethyl 2-oxopropanedioate
Molecular Formula
C7H10O5
InChI Key
DBKKFIIYQGGHJO-UHFFFAOYSA-N
SMILES
CCOC(=O)C(=O)C(=O)OCC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥94.0%
Refractive Index
1.4145-1.4205 @ 20?C

Description

Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl ketomalonate, reagent employed in Wittig and Aza-Wittig reactions for synthesis of triazoles, and 2-azadienes, respectively.

Solubility
Fully miscible with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text