Sodium 1-dodecanesulfonate, 99%
Sodium 1-dodecanesulfonate, 99%
Sodium 1-dodecanesulfonate, 99%
Sodium 1-dodecanesulfonate, 99%
Thermo Scientific Chemicals

Sodium 1-dodecanesulfonate, 99%

CAS: 2386-53-0 | C12H25NaO3S | 272.379 g/mol
製品番号(カタログ番号) A12829.03
または、製品番号A12829-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2386-53-0
IUPAC Namesodium dodecane-1-sulfonate
Molecular FormulaC12H25NaO3S
InChI KeyDAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCS([O-])(=O)=O
さらに表示
Assay (unspecified)≥98.5%
Water Content (Karl Fischer Titration)≤1.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (unspecified)≥98.5%
さらに表示
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jin, Y.; Wang, L.; Wang, T.; Chen, P.; Bi, Y.; Yu, L. Aggregation behavior of dodecylsulfonate-based surface active ionic liquids in water. J. Mol. Liq. 2015, 212, 23-29.
  2. Khan, A.; Khan, A. A. P; Asiri, A. M.; Alamry, K. A. Preparation and Characterization of hybrid graphene oxide composite and its application in paracetamol microbiosensor. Polym. Compos. 2015, 36, (2), 221-228.