Thermo Scientific Chemicals

5-Chlorovanillic acid, tech. 90%, Thermo Scientific Chemicals

Catalog number: A12841.14
25 g, Each
Thermo Scientific Chemicals

5-Chlorovanillic acid, tech. 90%, Thermo Scientific Chemicals

Catalog number: A12841.14
25 g, Each
Quantity
Catalog number: A12841.14
also known as A12841-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62936-23-6
IUPAC Name
3-chloro-4-hydroxy-5-methoxybenzoic acid
Molecular Formula
C8H7ClO4
InChI Key
XBRYEHVBBMSSCG-UHFFFAOYSA-N
SMILES
COC1=CC(=CC(Cl)=C1O)C(O)=O
Assay (Aqueous acid-base Titration)
≥88.0 to ≤110.0%
Appearance (Color)
Pale yellow or pale cream to cream
Assay (Silylated GC)
≥88.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text