(Trimethylsilyl)acetylene, 98%
(Trimethylsilyl)acetylene, 98%
(Trimethylsilyl)acetylene, 98%
Thermo Scientific Chemicals

(Trimethylsilyl)acetylene, 98%

CAS: 1066-54-2 | C5H10Si | 98.22 g/mol
製品番号(カタログ番号)数量
A12856.06
または、製品番号A12856-06
5 g
製品番号(カタログ番号) A12856.06
または、製品番号A12856-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1066-54-2
IUPAC Nameethynyltrimethylsilane
Molecular FormulaC5H10Si
InChI KeyCWMFRHBXRUITQE-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#C
さらに表示
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.3875-1.3920 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
(Trimethylsilyl)acetylene is used in the preparation of trimethylsilanyl-propiolic acid ethyl ester by reacting with carbonochloridic acid ethyl ester as well as in the synthesis of iodoalkenes by radical addition of perfluoroalkyl iodides. It is used as a nucleophile in Friedel-Crafts type acylations and alkylations reactions, as a ligand in organometallic chemistry and an useful reagent in cycloaddition reactions. It acts as a precursor to lithium trimethylsilylacetylide It finds application in the synthesis of (±)-estrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Trimethylsilyl)acetylene is used in the preparation of trimethylsilanyl-propiolic acid ethyl ester by reacting with carbonochloridic acid ethyl ester as well as in the synthesis of iodoalkenes by radical addition of perfluoroalkyl iodides. It is used as a nucleophile in Friedel-Crafts type acylations and alkylations reactions, as a ligand in organometallic chemistry and an useful reagent in cycloaddition reactions. It acts as a precursor to lithium trimethylsilylacetylide It finds application in the synthesis of (±)-estrone.

Solubility
Miscible with organic solvents.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Pd catalyzed coupling with aryl bromides or iodides gives aryl TMS alkynes, from which the TMS group is readily cleaved by K2CO3 in MeOH: Synthesis, 627 (1980); J. Org. Chem., 46, 2280 (1981). Coupling with 1-bromo-2-nitrobenzene or ethyl 2-bromocarbanilate gives indole precursors. Other terminal alkynes lead to 2-substituted indoles: Heterocycles, 24, 31 (1986):
  2. Cross-coupling with aryl halides can also be catalyzed by Pd-C in the presence of Ph3P, CuI and Et3N: Synth. Commun., 20, 2059 (1990); for an example, see 2-Bromonaphthalene, A15300. Pd catalyzed coupling with quinolinol and isoquinolinol triflates has also been reported: Tetrahedron, 51, 3737 (1995). For coupling with vinyl halides catalyzed by Tetrakis(triphenyl phosphine) palladium(0) , 10548 in the presence of CuI as an example of a route to enynes, see: Tetrahedron Lett., 30, 6997 (1989); Org. Synth. Coll., 9, 117 (1998). CuCl-TMEDA catalyzed oxidative (Hay) coupling gives bis(TMS)-1,3-butadiyne: Org. Synth. Coll., 8, 63 (1993).
  3. For the synthesis of iodoalkenes by radical addition of perfluoroalkyl iodides, see: J. Org. Chem., 47, 2251 (1982).
  4. Das, S.; Samanta, S.; Ray, S.; Biswas, P. 3, 6-Di (pyridin-2-yl)-1, 2, 4, 5-tetrazine capped Pd (0) nanoparticles: a catalyst for copper-free Sonogashira coupling of aryl halides in aqueous medium. RSC Adv. 2015, 5 (92), 75263-75267.
  5. Wang, H.; Olesik, S. V. Development of a new separation media using ultra-thin glassy carbon film modified silica. J. Chromatogr. A 2015, 1379, 56-64.