Thermo Scientific Chemicals

4-Chloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A12879.0I
5000 g, Each
Thermo Scientific Chemicals

4-Chloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A12879.0I
5000 g, Each
Quantity
Catalog number: A12879.0I
also known as A12879-0I
Price (USD)
Quantity
-

Chemical Identifiers

CAS
106-47-8
IUPAC Name
4-chloroaniline
Molecular Formula
C6H6ClN
InChI Key
QSNSCYSYFYORTR-UHFFFAOYSA-N
SMILES
NC1=CC=C(Cl)C=C1
Appearance (Color)
White to cream to brown or pale yellow
Assay (GC)
>97.5%
Form
Crystals or crystalline powder or flakes or granules
Comment
May darken on storage

Description

4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroaniline is an important building block used in the chemical industry for the production of drugs and dyestuffs. Some benzodiazepine drugs use 4-chloroaniline in their manufacture.

Solubility
Soluble in water (2.6 g/Liter at 20°C ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, chloroformates and nitrous acid.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text