Ethyl 4-dimethylaminobenzoate, 99%
Ethyl 4-dimethylaminobenzoate, 99%
Ethyl 4-dimethylaminobenzoate, 99%
Thermo Scientific Chemicals

Ethyl 4-dimethylaminobenzoate, 99%

CAS: 10287-53-3 | C11H15NO2 | 193.246 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A12888.36
または、製品番号A12888-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10287-53-3
IUPAC Nameethyl 4-(dimethylamino)benzoate
Molecular FormulaC11H15NO2
InChI KeyFZUGPQWGEGAKET-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(C=C1)N(C)C
さらに表示
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)60-66°C
Ethyl 4-dimethylaminobenzoate is employed as a photoinititator in cell encapsulation applications. It is also useful for organic synthesis. Further, it is used in UV-curing coatings and inks for the initiation of photo polymerization of unsaturated prepolymers. In addition to this, it acts as a active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-dimethylaminobenzoate is employed as a photoinititator in cell encapsulation applications. It is also useful for organic synthesis. Further, it is used in UV-curing coatings and inks for the initiation of photo polymerization of unsaturated prepolymers. In addition to this, it acts as a active pharmaceutical ingredient intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with reducing agents, oxidizing agents, bases and acids.
RUO – Research Use Only

General References:

  1. Meng, Q. Y.; Xu, D.; Di, Y. Y.; Gao, Z. F.; Dou, J. M. Low-temperature heat capacities and thermodynamic properties of ethyl 4-dimethylaminobenzoate (C11H15NO2). J. Therm. Anal. Calorim. 2014, 119 (3), 1923-1930.
  2. Contreras, P. P.; Tyagi, P.; Agarwal, S. Low volume shrinkage of polymers by photopolymerization of 1,1-bis(ethoxycarbonyl)-2-vinylcyclopropanes. Polym. Chem. 2015, 6 (12), 2297-2304.