(3-Isocyanatopropyl)triethoxysilane, 95%
(3-Isocyanatopropyl)triethoxysilane, 95%
(3-Isocyanatopropyl)triethoxysilane, 95%
Thermo Scientific Chemicals

(3-Isocyanatopropyl)triethoxysilane, 95%

CAS: 24801-88-5 | C10H21NO4Si | 247.366 g/mol
製品番号(カタログ番号) A12890.14
または、製品番号A12890-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS24801-88-5
IUPAC Nametriethoxy(3-isocyanatopropyl)silane
Molecular FormulaC10H21NO4Si
InChI KeyFRGPKMWIYVTFIQ-UHFFFAOYSA-N
SMILESCCO[Si](CCCN=C=O)(OCC)OCC
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥92.0%
Identification (FTIR)Conforms
Refractive Index1.4190-1.4240 @ 20°C
It finds its application as a component in hybrid organic/inorganic urethanes. It is also used in the synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a component in hybrid organic/inorganic urethanes. It is also used in the synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Kang Wook Lee and Thomas J. McCarthy. Synthesis of a polymer surface containing covalently attached triethoxysilane functionality: adhesion to glass.Macromolecules.,1988,21(12), 3353-3356.
  2. B. Wang, et al. New high-refractive-index organic/inorganic hybrid materials from sol-gel processing.Macromolecules.,1991,24(11), 3449-3450.