1,1,2,2-Tetrabromoethane, 97%
1,1,2,2-Tetrabromoethane, 97%
1,1,2,2-Tetrabromoethane, 97%
Thermo Scientific Chemicals

1,1,2,2-Tetrabromoethane, 97%

CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol
数量:
50 mL
250 mL
1000 mL
製品番号(カタログ番号) A12943.AK
または、製品番号A12943-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79-27-6
IUPAC Name1,1,2,2-tetrabromoethane
Molecular FormulaC2H2Br4
InChI KeyQXSZNDIIPUOQMB-UHFFFAOYSA-N
SMILESBrC(Br)C(Br)Br
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.6340-1.6390 @ 20*C
1,1,2,2-Tetrabromoethane is useful for the separation of minerals from supporting rock. It is used as a solvent for fats, greases, oils and waxes. It acts as a mercury substitute in the production of gauges and balancing equipment. It finds application as a floating agent used in processing and separation of mineral oils. Further, it is used as refractive index liquid in microscopy. In addition to this, it is also employed as a flame-retardant additive for polystyrenes, polyurethanes and polyolefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,2,2-Tetrabromoethane is useful for the separation of minerals from supporting rock. It is used as a solvent for fats, greases, oils and waxes. It acts as a mercury substitute in the production of gauges and balancing equipment. It finds application as a floating agent used in processing and separation of mineral oils. Further, it is used as refractive index liquid in microscopy. In addition to this, it is also employed as a flame-retardant additive for polystyrenes, polyurethanes and polyolefins.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, aluminum, magnesium, alkali metals and zinc.
RUO – Research Use Only

General References:

  1. Petrov, A. R.; Derheim, A.; Oetzel, J.; Leibold, M.; Bruhn, C.; Scheerer, S.; Oßwald, S.; Winter, R. F.; Siemeling, U. A Stable Planar-Chiral N-Heterocyclic Carbene with a 1,1'-Ferrocenediyl Backbone. Inorg. Chem. 2015, 54 (13), 6657-6670.
  2. Afreh, I.; Wujcik, E. K.; Blasdel, N.; Sauer, B.; Kaya, S.; Duirk, S.; Monty, C. N. Detection of halogenated organics by their inhibitory action in a catalytic reaction between dimethyl acetylenedicarboxylate and 2-methyl-4-nitroaniline. J. Anal. Chem. 2015, 70 (7), 825-830.