Thermo Scientific Chemicals

4-Fluorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A13028.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A13028.14
25 g, Each
Quantity
Catalog number: A13028.14
also known as A13028-14
Price (USD)
Price: 85.10
Online price: 73.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1194-02-1
IUPAC Name
4-fluorobenzonitrile
Molecular Formula
C7H4FN
InChI Key
AEKVBBNGWBBYLL-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)C#N
Form
Crystals or powder or crystalline powder or fused solid or irregular pieces
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to yellow
Assay (GC)
≥98.5%

Description

4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzonitrile is used as chemical intermediate, solvent for perfumes and pharmaceuticals, stabilizer for chlorinated solvents, HPLC analysis, catalyst and component of transition-metal complex catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text