Thermo Scientific Chemicals

3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A13043.14
25 g, Each
Thermo Scientific Chemicals

3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals

Catalog number: A13043.14
25 g, Each
Quantity
Catalog number: A13043.14
also known as A13043-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
151-18-8
IUPAC Name
3-aminopropanenitrile
Molecular Formula
C3H6N2
InChI Key
AGSPXMVUFBBBMO-UHFFFAOYSA-N
SMILES
NCCC#N
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4355-1.4395 @ 20*C

Description

3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

Solubility
Soluble in water.

Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text