5-Decyne, 98%
5-Decyne, 98%
5-Decyne, 98%
Thermo Scientific Chemicals

5-Decyne, 98%

CAS: 1942-46-7 | C10H18 | 138.254 g/mol
製品番号(カタログ番号) A13050.09
または、製品番号A13050-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1942-46-7
IUPAC Namedec-5-yne
Molecular FormulaC10H18
InChI KeyJWBQJUFCNOLNNC-UHFFFAOYSA-N
SMILESCCCCC#CCCCC
さらに表示
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4315-1.4355 @ 20?C
FormLiquid
5-Decyne is used as APIs and intermediates. It is also used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Decyne is used as APIs and intermediates. It is also used for chemical research.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Toshio Masuda; Yoshinori Kuwane; Toshinobu Higashimura. Polymerization of symmetrical dialkylacetylenes (3-Hexyne, 4-Octyne, and 5-Decyne) by WCl6·Ph4Sn and MoCl5·Ph4Sn. Polymer Journal. 1981, 13, (3),301-303