Thermo Scientific Chemicals

3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Catalog number: A13120.14
25 g, Each
Thermo Scientific Chemicals

3-Mercapto-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Catalog number: A13120.14
25 g, Each
Quantity
Catalog number: A13120.14
also known as A13120-14
Price (USD)
Price: 151.00
Online price: 131.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3179-31-5
IUPAC Name
2,3-dihydro-1H-1,2,4-triazole-3-thione
Molecular Formula
C2H3N3S
InChI Key
AFBBKYQYNPNMAT-UHFFFAOYSA-N
SMILES
S=C1NNC=N1
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%

Description

3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.

Solubility
Soluble in hot water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text