1-Chloro-3-fluorobenzene, 99%
1-Chloro-3-fluorobenzene, 99%
1-Chloro-3-fluorobenzene, 99%
Thermo Scientific Chemicals

1-Chloro-3-fluorobenzene, 99%

CAS: 625-98-9 | C6H4ClF | 130.55 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13123.14
または、製品番号A13123-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS625-98-9
Identification (FTIR)Conforms
Refractive Index1.4925-1.4965 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.Onda; T.Odaka; H.Miyazaki; M. Mori; I.Yamaguchi; Y.Niide. Microwave Spectrum, Quadrupole Coupling Constant Tensors, and Dipole Moments of 1-Chloro-3-Fluorobenzene. Journal of Molecular Spectroscopy. 1994, 165, (2),426-432
  2. Antonio Ramirez; John Candler; Crystal G.Bashore; Michael C.Wirtz, Jotham W.Coe; David B.Collum. Formation of benzynes from 2,6-dihaloaryllithiums: mechanistic basis of the regioselectivity. Journal of the American Chemical Society. 2004, 126, (45),14700-14701
  3. Can be lithiated with n-BuLi at the 2-position. Reaction of the metallated species with CO2 gives 2-chloro-6-fluorobenzoic acid in 93% yield: Tetrahedron Lett., 37, 6551 (1996).