1,2-Phenylenediacetonitrile, 98%
1,2-Phenylenediacetonitrile, 98%
1,2-Phenylenediacetonitrile, 98%
Thermo Scientific Chemicals

1,2-Phenylenediacetonitrile, 98%

CAS: 613-73-0 | C10H8N2 | 156.19 g/mol
製品番号(カタログ番号) A13142.18
または、製品番号A13142-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS613-73-0
IUPAC Name2-[2-(cyanomethyl)phenyl]acetonitrile
Molecular FormulaC10H8N2
InChI KeyFWPFXBANOKKNBR-UHFFFAOYSA-N
SMILESN#CCC1=CC=CC=C1CC#N
さらに表示
Assay (GC)≥97.5%
Melting Point (clear melt)55.5-61.5?C
Appearance (Color)White to pale yellow to pale brown
FormCrystals or powder or crystalline powder
1,2-Phenylenediacetonitrile is used as API and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Phenylenediacetonitrile is used as API and intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Claudia Caltagirone; Philip A.Gale. Anion receptor chemistry: highlights from 2007. Chemical Society Reviews. 2009, 38, (2),520-563
  2. Rajesh Mathur; Pavan Mathur. Monomeric Cu (II) complexes with tetradentate bis (benzimidazole) ligand ; synthesis, EPR, spectral and electrochemical studies. Polyhedron. 1998, 17, (16),2607-2615