Hexamethyldisilane, 98+%
Hexamethyldisilane, 98+%
Hexamethyldisilane, 98+%
Thermo Scientific Chemicals

Hexamethyldisilane, 98+%

CAS: 1450-14-2 | C6H18Si2 | 146.38 g/mol
製品番号(カタログ番号) A13155.14
または、製品番号A13155-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1450-14-2
IUPAC Namehexamethyldisilane
Molecular FormulaC6H18Si2
InChI KeyNEXSMEBSBIABKL-UHFFFAOYSA-N
SMILESC[Si](C)(C)[Si](C)(C)C
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.4200-1.4240 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilane acts as a silylating reagent for allylic acetates, aryl halides and diketones. It is a source material for vapor deposition during silicon carbide growth. It is also used in electronic and semiconducter industries.

Solubility
Insoluble in water. Soluble in alcohol, ether and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, acids and bases.
RUO – Research Use Only

General References:

  1. B.Beagley; J.J.Monaghan; T.G.Hewitt. Electron-diffraction studies of tetramethylsilane and hexamethyldisilane, and discussion of the lengths of Si-C bonds. Journal of Molecular Structure. 1971, 8, (4),401-411 
  2. M.Simon; T.Lebrun; R.Martins; G.G.B. de Souza; I.Nenner; M.Lavollee; P.Morin. Multicoincidence mass spectrometry applied to hexamethyldisilane excited around the silicon 2p edge. J. Phys. Chem. 1993, 97, (20),5228-5237
  3. Precursor of TMSLi, TMSNa and TMSK by cleavage with alkyllithiums or alkoxides. Reaction of the metallated derivatives with aryl halides gives aryltrimethylsilanes: J. Org. Chem., 42, 2654 (1977). The anion adds 1,4-to ɑß-enones to give ß-silyl ketones: J. Org. Chem., 41, 3063 (1976); Tetrahedron Lett., 24, 3497 (1983). The anion may also be used to deoxygenate secondary nitroalkanes (to ketoximes), nitrones (to imines), and heterocyclic N-oxides: J. Org. Chem., 64, 2211 (1999).
  4. Also cleaved by TBAF to give the salt-free silyl anion, which adds to aldehydes to give, after acid hydrolysis, ɑ-hydroxy silanes: J. Org. Chem., 48, 912 (1983). TBAF also catalyzes the silylation of OH groups under very mild conditions: Tetrahedron Lett., 35, 8413 (1994). See Appendix 4.
  5. Couples with aryl, benzyl or allyl halides in the presence of Tetrakis(triphenyl phosphine) palladium(0) , 10548, to give the corresponding silanes: J. Organomet. Chem., 148, 97 (1978); 225, 331 (1982).
  6. Aryl and alkenyl nitriles undergo ipso-silylation-decyanation, catalyzed by Chloro(1,5-cyclooctadiene) rhodium(I) dimer, 10466, to give the corresponding trimethylsiliyl derivatives: J. Am. Chem. Soc., 128, 8152 (2006).