Adipoyl chloride, 98%
Adipoyl chloride, 98%
Adipoyl chloride, 98%
Thermo Scientific Chemicals

Adipoyl chloride, 98%

CAS: 111-50-2 | C6H8Cl2O2 | 183.028 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13168.14
または、製品番号A13168-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS111-50-2
IUPAC Namehexanedioyl dichloride
Molecular FormulaC6H8Cl2O2
InChI KeyPWAXUOGZOSVGBO-UHFFFAOYSA-N
SMILESClC(=O)CCCCC(Cl)=O
さらに表示
Appearance (Color)Clear colorless to yellow to brown to dark brown
Assay (GC)≥97.5%
Refractive Index1.4670-1.4730 @ 20?C
FormLiquid
CommentMay darken on storage
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.

Solubility
Immiscible with water.

Notes
Incompatible strong oxidizing agents, bases and acids.
RUO – Research Use Only

General References:

  1. Al Tamimi, I. O.; Al Heeti, M. I.; Aziz, J. Z. Preparation of Dioxadiazole from Aryl Acid Hydrazide with Adipoyl Chloride. Int. J. Curr. Microbiol. App. Sci. 2015, 4 (12), 630-639.
  2. Dasgupta, Q.; Chatterjee, K.; Madras, G. Controlled Release of Salicylic Acid from Biodegradable Cross-Linked Polyesters. Mol. Pharmaceutics 2015, 12 (9), 3479-3489.