Thermo Scientific Chemicals

Naphthalene, 99+%, Thermo Scientific Chemicals

Catalog number: A13188.0I
5000 g, Each
Thermo Scientific Chemicals

Naphthalene, 99+%, Thermo Scientific Chemicals

Catalog number: A13188.0I
5000 g, Each
Quantity
Catalog number: A13188.0I
also known as A13188-0I
Price (USD)
Price: 172.00
Online price: 146.65
Your price:
Quantity
-

Chemical Identifiers

CAS
91-20-3
IUPAC Name
naphthalene
Molecular Formula
C10H8
InChI Key
UFWIBTONFRDIAS-UHFFFAOYSA-N
SMILES
C1=CC2=CC=CC=C2C=C1
Form
Crystals or powder or crystalline powder or fused solid or flakes
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (GC)
≥99.0%
Melting Point (clear melt)
78.0-84.0?C

Description

Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.

Solubility
Soluble in benzene, toluene, chloroform, carbon disulfide, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text