3-Aminopyridine, 99%
3-Aminopyridine, 99%
3-Aminopyridine, 99%
3-Aminopyridine, 99%
Thermo Scientific Chemicals

3-Aminopyridine, 99%

CAS: 462-08-8 | C5H6N2 | 94.12 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A13216.0B
または、製品番号A13216-0B
価格(JPY)
-
見積もりを依頼する
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS462-08-8
IUPAC Namepyridin-3-amine
Molecular FormulaC5H6N2
InChI KeyCUYKNJBYIJFRCU-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1
さらに表示
Melting Point (clear melt)55-63°C
Appearance (Color)White to cream to brown or orange
FormCrystals or crystalline powder or lumps or fused solid or flakes
Assay (GC)≥98.5%
Water Content (Karl Fischer Titration)<0.6%
さらに表示
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

Solubility
It is soluble in water as well as soluble in alcohol, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
RUO – Research Use Only

General References:

  1. RA Finch, MC Liu, SP Grill, WC Rose, R LoomisTriapine (3-aminopyridine-2-carboxaldehyde-thiosemicarbazone): a potent inhibitor of ribonucleotide reductase activity with broad spectrum antitumor activity. Biochemical Pharmacology. 2000, 59,(8), 983-991.
  2. S Caron.; SS Masset.; DE Bogle. An efficient and cost-effective synthesis of 2-phenyl-3-aminopyridine Org. Proc. Res. Dev.2001, 5,(3), 254-256.