4-Methylphenoxyacetic acid, 98%
4-Methylphenoxyacetic acid, 98%
4-Methylphenoxyacetic acid, 98%
4-Methylphenoxyacetic acid, 98%
Thermo Scientific Chemicals

4-Methylphenoxyacetic acid, 98%

CAS: 940-64-7 | C9H10O3 | 166.176 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A13230.09
または、製品番号A13230-09
価格(JPY)
-
見積もりを依頼する
数量:
10 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン2047591
CAS940-64-7
化学物質名または材質4-Methylphenoxyacetic acid
EINECS番号213-374-7
式量166.18
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only