2'-Methylacetanilide, 98+%
2'-Methylacetanilide, 98+%
2'-Methylacetanilide, 98+%
Thermo Scientific Chemicals

2'-Methylacetanilide, 98+%

CAS: 120-66-1 | C9H11NO | 149.193 g/mol
製品番号(カタログ番号) A13235.14
または、製品番号A13235-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-66-1
IUPAC NameN-(2-methylphenyl)acetamide
Molecular FormulaC9H11NO
InChI KeyBPEXTIMJLDWDTL-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=CC=C1C
さらに表示
Melting Point (clear melt)108-113?C
Appearance (Color)White to cream or pale pink
Assay (GC)≥98.0%
FormPowder
Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetanilide is used as an inhibitor of peroxides and stabilizer for cellulose ester varnishes. It is used as an intermediate for the synthesis of rubber accelerators, dyes and dye intermediate and camphor. It is used as a precursor in penicillin synthesis and other pharmaceuticals including painkillers and intermediates. 

Solubility
It is slightly soluble in acetone, chloroform, toluene, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Stable under ordinary conditions.
RUO – Research Use Only

General References:

  1. H Akhavan-Tafti.; R DeSilva.; Z Arghavani. Characterization of acridancarboxylic acid derivatives as chemiluminescent peroxidase substrates. J. Org. Chem. 1998, 63,(4), 930-937.
  2. JR Hanson.; ICR Medina. Oxidative Iodination of Aromatic Amides Using Sodium Perborate or Hydrogen Peroxide with Sodium Tungstate. 1998, 63,(4), 204-205.