Thermo Scientific Chemicals

3-Chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13254.36
500 g, Each
Thermo Scientific Chemicals

3-Chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13254.36
500 g, Each
Quantity
Catalog number: A13254.36
also known as A13254-36
Price (USD)
Price: 675.00
Online price: 589.65
Your price:
Quantity
-

Chemical Identifiers

CAS
587-04-2
IUPAC Name
3-chlorobenzaldehyde
Molecular Formula
C7H5ClO
InChI Key
SRWILAKSARHZPR-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(C=O)=C1
Form
Liquid
Refractive Index
1.5615-1.5665 @ 20?C
Free acid (titration)
≤3%
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%

Description

3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.

Solubility
It is soluble in alcohol, ether, and acetone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong bases, Strong oxidizing agents, Strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text