3-Methylthiophene-2-carbonitrile, 96%
3-Methylthiophene-2-carbonitrile, 96%
3-Methylthiophene-2-carbonitrile, 96%
3-Methylthiophene-2-carbonitrile, 96%
Thermo Scientific Chemicals

3-Methylthiophene-2-carbonitrile, 96%

CAS: 55406-13-8 | C6H5NS | 123.173 g/mol
製品番号(カタログ番号) A13305.06
または、製品番号A13305-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS55406-13-8
IUPAC Name3-methylthiophene-2-carbonitrile
Molecular FormulaC6H5NS
InChI KeyALZHYEITUZEZMT-UHFFFAOYSA-N
SMILESCC1=C(SC=C1)C#N
さらに表示
Refractive Index1.5505-1.5555 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only