1,4-Diethylbenzene, 98%
1,4-Diethylbenzene, 98%
1,4-Diethylbenzene, 98%
Thermo Scientific Chemicals

1,4-Diethylbenzene, 98%

CAS: 105-05-5 | C10H14 | 134.22 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A13343.14
または、製品番号A13343-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS105-05-5
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4930-1.4970 @ 20°C
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.

Solubility
It is insoluble in water. But soluble in ethanol, benzene,carbon tetrachloride.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. SN Rogak.; U Pott.; T Dann.; D Wang. Gaseous emissions from vehicles in a traffic tunnel in Vancouver, British Columbia. Journal of the Air & Waste Management Association. 1998, 48,(7), 604-615.