Thermo Scientific Chemicals

Tetrahydropyran, 98+%, Thermo Scientific Chemicals

Catalog number: A13392.AP
500 mL, Each
Thermo Scientific Chemicals

Tetrahydropyran, 98+%, Thermo Scientific Chemicals

Catalog number: A13392.AP
500 mL, Each
Quantity
Catalog number: A13392.AP
also known as A13392-AP
Price (USD)
Price: 206.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
142-68-7
IUPAC Name
oxane
Molecular Formula
C5H10O
InChI Key
DHXVGJBLRPWPCS-UHFFFAOYSA-N
SMILES
C1CCOCC1
Refractive Index
1.4190-1.4230 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Oxidizing agents, Strong acids. May form peroxides in the absence of inhibitors. Light sensitive
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text